Assay Details
Gene Symbol: Cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053527.1 | NP_445979.1 | 15 - 16 | 2404 | 67 | |
GenBank mRNA | AF000578.2 | - | 15 - 16 | 2404 | 67 | |
BC087007.1 | - | 16 - 17 | 2451 | 67 | ||
BC161839.1 | - | 15 - 16 | 2368 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint signaling
G2/M transition of mitotic cell cycle
DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
DNA binding
chromatin binding
RNA binding
protein binding
protein domain specific binding
WD40-repeat domain binding
DNA damage checkpoint signaling
G2/M transition of mitotic cell cycle
DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
DNA binding
chromatin binding
RNA binding
protein binding
protein domain specific binding
WD40-repeat domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P332817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169197 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P332817 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P332817 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P332817 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P332817 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101517 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56608 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13491 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104928 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10921882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921888 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921884 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921885 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921893 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921895 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921890 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921887 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921892 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921883 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921881 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921891 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921894 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921889 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921886 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921896 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10921880 | - | - |
Rat Genome 230 2.0 Array | 1398793_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398793_at | - | - |
Affymetrix HT Rat Focus Array | 1398793_at | - | - |
Affymetrix HT Rat Focus Array | 1398793_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0900002426.rn.1 | TC0900000256.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900002425.rn.1 | TC0900000256.rn.1 | RTA TC |
Rat Expression Set 230 | 1398793_PM_at | - | - |
Rat Expression Set 230 | 1398793_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921880 | - | - |
Rat Genome U34 Set | AF000578_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21067856 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373826 | - | - |
RatRef-12 Expression BeadChip | ILMN_48428 | - | - |
