Assay Details
Gene Symbol: F11r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053796.1 | NP_446248.1 | 3 - 4 | 309 | 57 | |
GenBank mRNA | AF276998.1 | - | 3 - 4 | 309 | 57 | |
BC065309.1 | - | 3 - 4 | 291 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
