Assay Details
Gene Symbol: Opcml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053848.1 | NP_446300.1 | 5 - 6 | 1255 | 73 | |
XM_008766041.1 | XP_008764263.1 | 3 - 4 | 413 | 73 | ||
GenBank mRNA | M88709.1 | - | 1405 | 73 | ||
M88710.1 | - | 5 - 6 | 1086 | 73 | ||
M88711.1 | - | 5 - 6 | 1255 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
