Assay Details
Gene Symbol: Pdia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053849.1 | NP_446301.1 | 8 - 9 | 1568 | 128 | |
GenBank mRNA | BC061535.1 | - | 8 - 9 | 1382 | 128 | |
M86870.1 | - | 8 - 9 | 1568 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
