Assay Details
Gene Symbol: Vcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053864.2 | NP_446316.1 | 3 - 4 | 517 | 78 | |
GenBank mRNA | BC060518.1 | - | 3 - 4 | 517 | 78 | |
U11760.1 | - | 3 - 4 | 549 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
