Assay Details
Gene Symbol: Cdc123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053877.3 | NP_446329.1 | 3 - 4 | 243 | 61 | |
GenBank mRNA | BC061527.1 | - | 3 - 4 | 244 | 61 | |
U34843.1 | - | 3 - 4 | 255 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
