Assay Details
Gene Symbol: Chst10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080397.1 | NP_536322.1 | 4 - 5 | 687 | 72 | |
XM_006244730.2 | XP_006244792.1 | 5 - 6 | 836 | 72 | ||
XM_006244732.2 | XP_006244794.1 | 4 - 5 | 494 | 72 | ||
XM_008766985.1 | XP_008765207.1 | 4 - 5 | 618 | 72 | ||
XM_008766987.1 | XP_008765209.1 | 4 - 5 | 588 | 72 | ||
GenBank mRNA | AF022729.1 | - | 4 - 5 | 687 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
