Assay Details
Gene Symbol: Gpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022215.2 | NP_071551.2 | 3 - 4 | 378 | 95 | |
XM_006257376.1 | XP_006257438.1 | 4 - 5 | 580 | 95 | ||
GenBank mRNA | AB002558.1 | - | 3 - 4 | 366 | 95 | |
BC088396.1 | - | 3 - 4 | 378 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
