Assay Details
Gene Symbol: Asic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022234.2 | NP_071570.2 | 8 - 9 | 1709 | 63 | |
XM_006245275.2 | XP_006245337.1 | 9 - 10 | 1838 | 63 | ||
GenBank mRNA | AJ242554.1 | - | 8 - 9 | 1690 | 63 | |
AJ271642.1 | - | 8 - 9 | 1613 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
