Assay Details
Gene Symbol: Birc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022274.1 | NP_071610.1 | 1 - 2 | 118 | 70 | |
GenBank mRNA | AF276775.1 | - | 1 - 2 | 118 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
