Assay Details
Gene Symbol: Prdx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022540.1 | NP_071985.1 | 4 - 5 | 480 | 74 | |
GenBank mRNA | AF106944.1 | - | 4 - 5 | 480 | 74 | |
BC060567.1 | - | 4 - 5 | 481 | 74 | ||
FQ212826.1 | - | 4 - 5 | 460 | 74 | ||
FQ219655.1 | - | 4 - 5 | 477 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
