Assay Details
Gene Symbol: Pgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022847.1 | NP_074038.1 | 6 - 7 | 2321 | 100 | |
GenBank mRNA | HM037361.1 | - | 2873 | 100 | ||
HM037362.1 | - | 2873 | 100 | |||
L16922.1 | - | 6 - 7 | 2321 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
