Assay Details
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270838.1 | NP_001257767.1 | 4 - 5 | 587 | 60 | |
NM_001270839.1 | NP_001257768.1 | 4 - 5 | 587 | 60 | ||
NM_001270840.1 | NP_001257769.1 | 4 - 5 | 587 | 60 | ||
NM_022941.4 | NP_075230.1 | 4 - 5 | 587 | 60 | ||
XM_006250261.2 | XP_006250323.1 | 4 - 5 | 696 | 60 | ||
XM_008769758.1 | XP_008767980.1 | 4 - 5 | 699 | 60 | ||
XM_008769759.1 | XP_008767981.1 | 4 - 5 | 699 | 60 | ||
GenBank mRNA | AB104736.1 | - | 4 - 5 | 640 | 60 | |
AB105072.1 | - | 4 - 5 | 640 | 60 | ||
AB204897.1 | - | 3 - 4 | 438 | 60 | ||
AB204898.1 | - | 3 - 4 | 438 | 60 | ||
AB204899.1 | - | 3 - 4 | 438 | 60 | ||
AB204900.1 | - | 3 - 4 | 438 | 60 | ||
AF133094.1 | - | 3 - 4 | 438 | 60 | ||
AF133095.1 | - | 3 - 4 | 438 | 60 | ||
AF233431.1 | - | 1 - 2 | 91 | 60 | ||
BC089763.1 | - | 4 - 5 | 587 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
