Assay Details
Gene Symbol: Mrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024001.1 | NP_076491.1 | 4 - 5 | 458 | 114 | |
XM_006253937.2 | XP_006253999.1 | 4 - 5 | 576 | 114 | ||
GenBank mRNA | AF288289.1 | - | 4 - 5 | 458 | 114 | |
BC083554.1 | - | 4 - 5 | 434 | 114 | ||
BC098916.1 | - | 4 - 5 | 433 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
