Assay Details
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111098.1 | NP_001104568.1 | 5 - 6 | 545 | 75 | |
NM_030857.2 | NP_110484.1 | 5 - 6 | 608 | 75 | ||
XM_006237834.2 | XP_006237896.1 | 5 - 6 | 924 | 75 | ||
GenBank mRNA | AF000300.1 | - | 5 - 6 | 453 | 75 | |
AF000301.1 | - | 5 - 6 | 545 | 75 | ||
AF000302.1 | - | 5 - 6 | 543 | 75 | ||
L14782.1 | - | 4 - 5 | 402 | 75 | ||
L14823.1 | - | 4 - 5 | 339 | 75 | ||
L14951.1 | - | 5 - 6 | 611 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
