Assay Details
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111098.1 | NP_001104568.1 | 9 - 10 | 1142 | 73 | |
NM_030857.2 | NP_110484.1 | 9 - 10 | 1205 | 73 | ||
XM_006237834.2 | XP_006237896.1 | 9 - 10 | 1521 | 73 | ||
GenBank mRNA | AF000300.1 | - | 9 - 10 | 1050 | 73 | |
AF000301.1 | - | 9 - 10 | 1142 | 73 | ||
AF000302.1 | - | 9 - 10 | 1140 | 73 | ||
L14951.1 | - | 9 - 10 | 1208 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
