Assay Details
Gene Symbol: Dync1li2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031026.1 | NP_112288.1 | 4 - 5 | 527 | 105 | |
XM_006255053.2 | XP_006255115.1 | 4 - 5 | 741 | 105 | ||
XM_006255054.2 | XP_006255116.1 | 4 - 5 | 741 | 105 | ||
GenBank mRNA | BC089794.1 | - | 4 - 5 | 548 | 105 | |
U15138.1 | - | 4 - 5 | 527 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
