Assay Details
Gene Symbol: Itpka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031045.2 | NP_112307.2 | 5 - 6 | 1147 | 64 | |
GenBank mRNA | M29787.1 | - | 5 - 6 | 1147 | 64 | |
X56917.1 | - | 5 - 6 | 1138 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
