Assay Details
Gene Symbol: Mlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031053.1 | NP_112315.1 | 7 - 8 | 595 | 93 | |
XM_008766652.1 | XP_008764874.1 | 5 - 6 | 357 | 93 | ||
GenBank mRNA | U80054.1 | - | 7 - 8 | 595 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
