Assay Details
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031056.1 | NP_112318.1 | 3 - 4 | 544 | 71 | |
GenBank mRNA | BC072509.1 | - | 3 - 4 | 575 | 71 | |
X83537.1 | - | 3 - 4 | 553 | 71 | ||
X91785.1 | - | 3 - 4 | 544 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
