Assay Details
Gene Symbol: Tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031131.1 | NP_112393.1 | 5 - 6 | 1145 | 113 | |
XM_006250448.2 | XP_006250510.1 | 4 - 5 | 1971 | 113 | ||
GenBank mRNA | AF135598.1 | - | 4 - 5 | 755 | 113 | |
AF153012.2 | - | 5 - 6 | 1145 | 113 | ||
AF153013.1 | - | 4 - 5 | 1061 | 113 | ||
AJ132718.1 | - | 4 - 5 | 769 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
