Assay Details
Gene Symbol: Slc7a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031341.1 | NP_112631.1 | 3 - 4 | 832 | 67 | |
XM_006252020.2 | XP_006252082.1 | 3 - 4 | 768 | 67 | ||
XM_006252021.2 | XP_006252083.1 | 6 - 7 | 1237 | 67 | ||
XM_006252023.2 | XP_006252085.1 | 3 - 4 | 773 | 67 | ||
XM_006252024.2 | XP_006252086.1 | 3 - 4 | 1462 | 67 | ||
XM_008770705.1 | XP_008768927.1 | 5 - 6 | 1180 | 67 | ||
GenBank mRNA | AB020520.1 | - | 3 - 4 | 832 | 67 | |
AF200684.1 | - | 2 - 3 | 640 | 67 | ||
BC091142.1 | - | 3 - 4 | 747 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
