Assay Details
Gene Symbol: Il1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031512.2 | NP_113700.2 | 3 - 4 | 148 | 75 | |
GenBank mRNA | BC091141.1 | - | 3 - 4 | 148 | 75 | |
M98820.1 | - | 3 - 4 | 168 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
