Assay Details
Gene Symbol: Mybph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031813.1 | NP_114001.1 | 2 - 3 | 83 | ||
XM_006249888.2 | XP_006249950.1 | 2 - 3 | 488 | 82 | ||
XM_006249889.2 | XP_006249951.1 | 2 - 3 | 487 | 82 | ||
GenBank mRNA | AF077338.1 | - | 2 - 3 | 83 | ||
BC061993.1 | - | 2 - 3 | 417 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
