Assay Details
Gene Symbol: Smarcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031983.2 | NP_114189.1 | 7 - 8 | 1002 | 83 | |
GenBank mRNA | AB003504.1 | - | 7 - 8 | 732 | 83 | |
BC062063.1 | - | 7 - 8 | 1002 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
