Assay Details
Gene Symbol: Pawr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033485.2 | NP_277020.1 | 4 - 5 | 866 | 100 | |
XM_006241317.2 | XP_006241379.1 | 5 - 6 | 1079 | 100 | ||
GenBank mRNA | U05989.1 | - | 4 - 5 | 866 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
