Assay Details
Gene Symbol: Angpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053546.1 | NP_445998.1 | 8 - 9 | 1620 | 60 | |
XM_006241609.2 | XP_006241671.1 | 8 - 9 | 1965 | 60 | ||
XM_008765463.1 | XP_008763685.1 | 8 - 9 | 1149 | 60 | ||
XM_008765464.1 | XP_008763686.1 | 8 - 9 | 3613 | 60 | ||
GenBank mRNA | AB080023.1 | - | 8 - 9 | 1620 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
ovarian follicle development
response to hypoxia
in utero embryonic development
negative regulation of protein phosphorylation
regulation of endothelial cell proliferation
sprouting angiogenesis
positive regulation of receptor internalization
negative regulation of cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
activation of transmembrane receptor protein tyrosine kinase activity
positive regulation of endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of skeletal muscle satellite cell proliferation
hemopoiesis
cell differentiation
heparin biosynthetic process
animal organ regeneration
positive regulation of protein ubiquitination
cell-substrate adhesion
regulation of tumor necrosis factor production
positive regulation of peptidyl-serine phosphorylation
response to vitamin B3
protein localization to cell surface
negative regulation of protein import into nucleus
negative regulation of apoptotic process
negative regulation of vascular permeability
regulation of I-kappaB kinase/NF-kappaB signaling
regulation of protein binding
negative regulation of neuron apoptotic process
positive regulation of blood vessel endothelial cell migration
response to estrogen
positive regulation of cell adhesion
Tie signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
protein homooligomerization
positive regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
glomerulus vasculature development
negative regulation of endothelial cell apoptotic process
regulation of macrophage migration inhibitory factor signaling pathway
molecular_function
signaling receptor binding
receptor tyrosine kinase binding
angiogenesis
ovarian follicle development
response to hypoxia
in utero embryonic development
negative regulation of protein phosphorylation
regulation of endothelial cell proliferation
sprouting angiogenesis
positive regulation of receptor internalization
negative regulation of cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
activation of transmembrane receptor protein tyrosine kinase activity
positive regulation of endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of skeletal muscle satellite cell proliferation
hemopoiesis
cell differentiation
heparin biosynthetic process
animal organ regeneration
positive regulation of protein ubiquitination
cell-substrate adhesion
regulation of tumor necrosis factor production
positive regulation of peptidyl-serine phosphorylation
response to vitamin B3
protein localization to cell surface
negative regulation of protein import into nucleus
negative regulation of apoptotic process
negative regulation of vascular permeability
regulation of I-kappaB kinase/NF-kappaB signaling
regulation of protein binding
negative regulation of neuron apoptotic process
positive regulation of blood vessel endothelial cell migration
response to estrogen
positive regulation of cell adhesion
Tie signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
protein homooligomerization
positive regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
glomerulus vasculature development
negative regulation of endothelial cell apoptotic process
regulation of macrophage migration inhibitory factor signaling pathway
molecular_function
signaling receptor binding
receptor tyrosine kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1058692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178950 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1058692 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1058692 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1058692 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1058692 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89945 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50179 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52499 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34549 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17014 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100537 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14749 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45207 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45921 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17832 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2491 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48123 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66554 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30530 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43483 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100611 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30798 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75973 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16575 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39461 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10903530 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903531 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903537 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903536 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903533 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903535 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903532 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903534 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10903529 | - | - |
Rat Genome 230 2.0 Array | 1381487_at | - | - |
Rat Genome 230 2.0 Array | 1381487_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381487_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381487_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700023655.rn.1 | TC0700003068.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10903529 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22299981 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364587 | - | - |
RatRef-12 Expression BeadChip | ILMN_64155 | - | - |
