Assay Details
Gene Symbol: Nox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053683.1 | NP_446135.1 | 3 - 4 | 379 | 78 | |
GenBank mRNA | AF152963.1 | - | 3 - 4 | 379 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
