Assay Details
Gene Symbol: Emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053719.1 | NP_446171.1 | 1 - 2 | 199 | 100 | |
GenBank mRNA | AJ009698.1 | - | 1 - 2 | 199 | 100 | |
BC061846.1 | - | 1 - 2 | 201 | 100 | ||
FQ213962.1 | - | 1 - 2 | 212 | 100 | ||
FQ228768.1 | - | 1 - 2 | 212 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
