Assay Details
Gene Symbol: Cuzd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054005.1 | NP_446457.1 | 2 - 3 | 488 | 89 | |
GenBank mRNA | AF022147.1 | - | 2 - 3 | 488 | 89 | |
AF167170.1 | - | 2 - 3 | 254 | 89 | ||
BC090345.1 | - | 2 - 3 | 234 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
