Assay Details
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054011.1 | NP_446463.2 | 3 - 4 | 340 | 73 | |
GenBank mRNA | AB027562.1 | - | 3 - 4 | 340 | 73 | |
FQ210473.1 | - | 3 - 4 | 467 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
