Assay Details
Gene Symbol: Echs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078623.2 | NP_511178.1 | 4 - 5 | 533 | 70 | |
GenBank mRNA | BC064655.1 | - | 4 - 5 | 533 | 70 | |
FQ218439.1 | - | 3 - 4 | 392 | 70 | ||
FQ218627.1 | - | 1 - 2 | 77 | 70 | ||
X15958.1 | - | 4 - 5 | 527 | 70 |
Gene Symbol: LOC100911186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748886.2 | XP_003748934.1 | 4 - 5 | 561 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
