Assay Details
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012832.3 | NP_036964.3 | 6 - 7 | 600 | 80 | |
XM_008759497.1 | XP_008757719.1 | 4 - 5 | 4458 | 80 | ||
XM_008759498.1 | XP_008757720.1 | 7 - 8 | 747 | 80 | ||
GenBank mRNA | AY574256.1 | - | 6 - 7 | 600 | 80 | |
AY671974.1 | - | 7 - 8 | 690 | 80 | ||
L31619.1 | - | 6 - 7 | 622 | 80 | ||
S53987.1 | - | 641 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
