Assay Details
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012881.2 | NP_037013.2 | 4 - 5 | 282 | 99 | |
XM_008769996.1 | XP_008768218.1 | 4 - 5 | 355 | 99 | ||
GenBank mRNA | AB001382.1 | - | 3 - 4 | 176 | 99 | |
BC078874.1 | - | 4 - 5 | 262 | 99 | ||
CV102664.1 | - | 4 - 5 | 282 | 99 | ||
FQ213473.1 | - | 4 - 5 | 271 | 99 | ||
FQ222192.1 | - | 4 - 5 | 298 | 99 | ||
M14656.1 | - | 4 - 5 | 255 | 99 | ||
M99252.1 | - | 4 - 5 | 291 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
