Assay Details
Gene Symbol: Anxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012904.2 | NP_037036.1 | 4 - 5 | 333 | 80 | |
XM_006231159.1 | XP_006231221.1 | 5 - 6 | 501 | 80 | ||
XM_008760295.1 | XP_008758517.1 | 4 - 5 | 420 | 80 | ||
GenBank mRNA | BC061710.1 | - | 4 - 5 | 331 | 80 | |
FQ213489.1 | - | 4 - 5 | 348 | 80 | ||
FQ215033.1 | - | 4 - 5 | 348 | 80 | ||
FQ216134.1 | - | 4 - 5 | 348 | 80 | ||
FQ228716.1 | - | 4 - 5 | 349 | 80 | ||
FQ229071.1 | - | 4 - 5 | 349 | 80 | ||
FQ229198.1 | - | 3 - 4 | 269 | 80 | ||
M19967.1 | - | 3 - 4 | 300 | 80 | ||
Y00446.1 | - | 4 - 5 | 334 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
