Assay Details
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012939.1 | NP_037071.1 | 1 - 2 | 86 | 87 | |
XM_008766440.1 | XP_008764662.1 | 1 - 2 | 137 | 87 | ||
GenBank mRNA | BC085352.1 | - | 1 - 2 | 101 | 87 | |
FQ210730.1 | - | 1 - 2 | 96 | 87 | ||
FQ219177.1 | - | 1 - 2 | 96 | 87 | ||
FQ219281.1 | - | 1 - 2 | 96 | 87 | ||
FQ219285.1 | - | 1 - 2 | 96 | 87 | ||
FQ219548.1 | - | 1 - 2 | 96 | 87 | ||
FQ219611.1 | - | 1 - 2 | 114 | 87 | ||
FQ223707.1 | - | 1 - 2 | 92 | 87 | ||
FQ228906.1 | - | 1 - 2 | 103 | 87 | ||
FQ228938.1 | - | 1 - 2 | 93 | 87 | ||
FQ228956.1 | - | 1 - 2 | 123 | 87 | ||
M38135.1 | - | 1 - 2 | 81 | 87 | ||
Y00708.1 | - | 1 - 2 | 86 | 87 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the M38135.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
