Assay Details
Gene Symbol: Adrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013108.2 | NP_037240.2 | 2 - 3 | 1453 | 90 | |
GenBank mRNA | M74716.1 | - | 2 - 3 | 1452 | 90 | |
S56481.1 | - | 2 - 3 | 1244 | 90 | ||
S73473.1 | - | 2 - 3 | 1283 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
