Assay Details
Gene Symbol: Il2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013195.1 | NP_037327.1 | 4 - 5 | 391 | 62 | |
XM_006241970.2 | XP_006242032.1 | 4 - 5 | 820 | 62 | ||
GenBank mRNA | M55050.1 | - | 4 - 5 | 391 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
