Assay Details
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013200.1 | NP_037332.1 | 13 - 14 | 1615 | 64 | |
XM_006242180.2 | XP_006242242.1 | 13 - 14 | 2071 | 64 | ||
GenBank mRNA | BC085761.1 | - | 13 - 14 | 1627 | 64 | |
D43623.1 | - | 12 - 13 | 1602 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
