Assay Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146061.1 | NP_001139533.1 | 8 - 9 | 1106 | 94 | |
NM_013214.4 | NP_037346.2 | 8 - 9 | 1034 | 94 | ||
XM_006239447.2 | XP_006239509.1 | 8 - 9 | 1134 | 94 | ||
XM_006239448.1 | XP_006239510.1 | 8 - 9 | 1042 | 94 | ||
GenBank mRNA | BC087716.1 | - | 8 - 9 | 986 | 94 | |
D88891.1 | - | 8 - 9 | 1106 | 94 | ||
FQ212652.1 | - | 5 - 6 | 585 | 94 | ||
FQ231740.1 | - | 5 - 6 | 585 | 94 | ||
Y09332.1 | - | 8 - 9 | 934 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid catabolic process
coenzyme A biosynthetic process
medium-chain fatty-acyl-CoA catabolic process
long-chain fatty-acyl-CoA catabolic process
medium-chain fatty acid biosynthetic process
palmitic acid biosynthetic process
fatty-acyl-CoA binding
palmitoyl-CoA hydrolase activity
long-chain fatty acyl-CoA binding
protein homodimerization activity
acyl-CoA hydrolase activity
carboxylic ester hydrolase activity
fatty acid catabolic process
coenzyme A biosynthetic process
medium-chain fatty-acyl-CoA catabolic process
long-chain fatty-acyl-CoA catabolic process
medium-chain fatty acid biosynthetic process
palmitic acid biosynthetic process
fatty-acyl-CoA binding
palmitoyl-CoA hydrolase activity
long-chain fatty acyl-CoA binding
protein homodimerization activity
acyl-CoA hydrolase activity
carboxylic ester hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P103921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61499 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P103921 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P103921 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P103921 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11753 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700500858 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59466 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19928 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22544 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76821 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44733 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83848 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49352 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700500858 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10874304 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874311 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874303 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874299 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874306 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874307 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874300 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874308 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874305 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874301 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874310 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10874298 | - | - |
Rat Genome 230 2.0 Array | 1388075_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370313_at | - | - |
Rat Genome 230 2.0 Array | 1370313_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388075_at | - | - |
Affymetrix HT Rat Focus Array | 1370313_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388075_at | - | - |
Affymetrix HT Rat Focus Array | 1388075_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370313_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500014999.rn.1 | TC0500001997.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500015000.rn.1 | TC0500001997.rn.1 | RTA TC |
Rat Expression Set 230 | 1388075_at | - | - |
Rat Expression Set 230 | 1388075_PM_at | - | - |
Rat Expression Set 230 | 1370313_at | - | - |
Rat Expression Set 230 | 1370313_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874298 | - | - |
Rat Genome U34 Set | U49694_at | - | - |
Rat Genome U34 Set | Y09332cds_s_at | - | - |
Rat Genome U34 Set | D88890_at | - | - |
Rat Genome U34 Set | Y09332cds_s_at | - | - |
Rat Genome U34 Set | D88890_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20757649 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366923 | - | - |
RatRef-12 Expression BeadChip | ILMN_52687 | - | - |
