Assay Details
Gene Symbol: Ptgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017232.3 | NP_058928.3 | 8 - 9 | 1258 | 85 | |
GenBank mRNA | AF233596.1 | - | 1382 | 85 | ||
L20085.1 | - | 8 - 9 | 1258 | 85 | ||
L25925.1 | - | 8 - 9 | 1264 | 85 | ||
S67722.1 | - | 1382 | 85 | |||
U03389.1 | - | 1382 | 85 | |||
U04300.1 | - | 8 - 9 | 1258 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
