Assay Details
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133316.1 | NP_579850.1 | 5 - 6 | 431 | 73 | |
XM_006238626.2 | XP_006238688.1 | 5 - 6 | 459 | 73 | ||
XM_006238627.2 | XP_006238689.1 | 5 - 6 | 470 | 73 | ||
GenBank mRNA | AF478683.1 | - | 5 - 6 | 431 | 73 | |
BC128728.1 | - | 5 - 6 | 361 | 73 | ||
FQ230684.1 | - | 4 - 5 | 538 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
