Assay Details
Gene Symbol: Ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133409.2 | NP_596900.1 | 11 - 12 | 1271 | 133 | |
GenBank mRNA | AF329194.1 | - | 11 - 12 | 1208 | 133 | |
BC062406.1 | - | 11 - 12 | 1271 | 133 | ||
FQ222624.1 | - | 1 - 2 | 101 | 133 | ||
FQ233927.1 | - | 5 - 6 | 598 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
