Assay Details
Gene Symbol: Il13ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133538.2 | NP_598222.2 | 5 - 6 | 586 | 91 | |
XM_006257427.2 | XP_006257489.1 | 4 - 5 | 454 | 91 | ||
XM_006257428.2 | XP_006257490.1 | 4 - 5 | 572 | 91 | ||
XM_006257430.2 | XP_006257492.1 | 3 - 4 | 385 | 91 | ||
XM_008773388.1 | XP_008771610.1 | 4 - 5 | 726 | 91 | ||
XM_008773389.1 | XP_008771611.1 | 5 - 6 | 613 | 91 | ||
GenBank mRNA | AF448818.1 | - | 5 - 6 | 586 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF448818.1, NM_133538.2, XM_006257427.2, XM_006257428.2, XM_006257430.2, XM_008773388.1, XM_008773389.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
