Assay Details
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085369.1 | NP_001078838.1 | 16 - 17 | 1754 | 73 | |
NM_134472.3 | NP_608302.2 | 15 - 16 | 1639 | 73 | ||
XM_006238632.2 | XP_006238694.1 | 16 - 17 | 1806 | 73 | ||
XM_006238633.1 | XP_006238695.1 | 15 - 16 | 1570 | 73 | ||
XM_006238634.2 | XP_006238696.1 | 15 - 16 | 2102 | 73 | ||
XM_006238635.2 | XP_006238697.1 | 16 - 17 | 1979 | 73 | ||
XM_006238636.2 | XP_006238698.1 | 16 - 17 | 1975 | 73 | ||
GenBank mRNA | CO403678.1 | - | 3 - 4 | 408 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
