Assay Details
Gene Symbol: Haus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138864.2 | NP_620219.2 | 4 - 5 | 490 | 61 | |
GenBank mRNA | AF092207.1 | - | 486 | 61 | ||
BC127484.1 | - | 4 - 5 | 490 | 61 |
Gene Symbol: NEWGENE_619909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772211.1 | XP_008770433.1 | 515 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
