Assay Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248989.2 | XP_006249051.1 | 11 - 12 | 3591 | 83 | |
GenBank mRNA | AF030243.1 | - | 6 - 7 | 795 | 83 | |
BP475867.1 | - | 1 - 2 | 83 | |||
FQ214452.1 | - | 8 - 9 | 1169 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
