Assay Details
Gene Symbol: Faim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144756.2 | NP_653357.1 | 3 - 4 | 390 | 84 | |
GenBank mRNA | AF044201.1 | - | 3 - 4 | 339 | 84 | |
BC087606.1 | - | 3 - 4 | 390 | 84 |
Gene Symbol: LOC102551901 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242293.2 | XP_006242355.1 | 3 - 4 | 616 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
