Assay Details
Gene Symbol: Ngfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012610.2 | NP_036742.2 | 5 - 6 | 1097 | 64 | |
GenBank mRNA | X05137.1 | - | 5 - 6 | 1097 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
