Assay Details
Gene Symbol: Slc9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012652.1 | NP_036784.1 | 9 - 10 | 2713 | 63 | |
GenBank mRNA | M85299.1 | - | 9 - 10 | 2713 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
